1,2-Di(4-pyridyl)ethylene 98%

Supplier: Ambeed
Warning

A197589-1G A197589-5G A197589-10G
103816-946EA 9 USD
103816-946 103847-566 77085-580 77085-582 77192-184 77192-186
1,2-Di(4-pyridyl)ethylene 98%
1,2-Di(4-pyridyl)ethylene

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂
Boiling Pt: 334.1 °C (760 mm Hg)
Melting Pt: 148…152 °C
Flash Pt: 126.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00006448
CAS Number: 13362-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR