4,4'-Dibromo-trans-stilbene 95%

Supplier: Ambeed
Danger

Synonyms: (E)-1,2-Bis(4-bromophenyl)ethene

A232124-1G A232124-5G A232124-10G A232124-250MG A232124-25G
103810-814EA 9 USD
103810-814 103850-568 77089-488 77089-490 77098-250 77098-252 77303-930
4,4'-Dibromo-trans-stilbene 95%
4,4'-Dibromo-trans-stilbene

Precaución: for Research Use Only

Formula: C₁₄H₁₀Br₂
Boiling Pt: 392.9 °C (760 mm Hg)
Melting Pt: 212 °C
Flash Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00448009
CAS Number: 18869-30-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR