(E)-1-(3-(2-(7-Chloroquinolin-2-yl)vinyl)phenyl)prop-2-en-1-ol 95%
Supplier: Ambeed
|
Warning
|
A328209-1G
A328209-5G
A328209-250MG
103825-496EA
55
USD
103825-496
103836-450
76839-412
76839-414
76989-216
(E)-1-(3-(2-(7-Chloroquinolin-2-yl)vinyl)phenyl)prop-2-en-1-ol 95%
(E)-1-(3-(2-(7-Chloroquinolin-2-yl)vinyl)phenyl)prop-2-en-1-ol
Precaución: for Research Use Only
Formula:
C₂₀H₁₆ClNO Boiling Pt: 504.1±38.0 °C (760 mm Hg) Melting Pt: 116…117.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09833496 CAS Number: 149968-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...