trans-β-Nitrostyrene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-(2-Nitrovinyl)benzene, trans-1-Nitro-2-phenylethylene
A117856-10G
A117856-25G
A117856-1G
A117856-5G
A117856-100G
103798-944EA
11
USD
103798-944
103824-836
77136-350
77136-352
77297-558
77311-618
77571-068
trans-β-Nitrostyrene 98%
trans-β-Nitrostyrene
Precaución: for Research Use Only
Formula:
C₈H₇NO₂ MW: 149.15 g/mol Boiling Pt: 255.8 °C (760 mm Hg) Melting Pt: 55…58 °C Flash Pt: 117.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007402 CAS Number: 5153-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...