Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide 97%

Supplier: Ambeed
Danger

Synonyms: (Diphenylphosphoryl)(mesityl)methanone

A117380-10G A117380-25G A117380-5G A117380-500G A117380-100G
103824-756EA 6 USD
103824-756 76870-066 76870-068 77311-534 77571-034 77610-962
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide 97%
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00192110
CAS Number: 75980-60-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR