α,α-Difluoroanisole 98%

Supplier: Ambeed
Danger

Synonyms: (Difluoromethoxy)benzene, alpha,alpha-Difluoroanisole

A513974-25G A513974-10G A513974-1G
103839-534EA 18 USD
103839-534 76843-516 77116-648 77116-650
α,α-Difluoroanisole 98%
α,α-Difluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MW: 144.1200 g/mol
Boiling Pt: 138 °C (760 mm Hg)
Flash Pt: 43.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00068184
CAS Number: 458-92-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR