(DHQ)2PYR 97%
Supplier: Ambeed
|
Warning
|
A159222-100MG
A159222-250MG
A159222-1G
103805-234EA
248
USD
103805-234
103843-450
77138-058
77138-060
77300-392
(DHQ)2PYR 97%
(DHQ)2PYR
Precaución: for Research Use Only
Formula:
C₅₆H₆₀N₆O₄ MW: 881.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00198109 CAS Number: 149820-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...