(Cyclopropylmethyl)triphenylphosphonium bromide 95%
Supplier: Ambeed
|
Warning
|
A182901-100G
A182901-10G
A182901-250MG
A182901-1G
A182901-25G
103815-580EA
9
USD
103815-580
103846-142
76720-960
76720-962
76720-964
77083-720
77083-722
(Cyclopropylmethyl)triphenylphosphonium bromide 95%
(Cyclopropylmethyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₁₄H₂₉ClNP MW: 397.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00051878 CAS Number: 14799-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...