(Cyanomethyl)triphenylphosphonium chloride 95%

Supplier: Ambeed
Warning

A102870-100G A102870-10G A102870-25G A102870-500G A102870-5G
103822-132EA 9 USD
103822-132 76687-094 76687-096 76687-098 76687-100 77283-008
(Cyanomethyl)triphenylphosphonium chloride 95%
(Cyanomethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₀H₁₇ClNP
MW: 337.79 g/mol
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD00031672
CAS Number: 4336-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR