4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 98%

Supplier: Ambeed
Warning

Synonyms: (Chloro(4-methoxyphenyl)methylene)dibenzene, Mmt-Cl

A656015-25G A656015-5G A656015-100G A656015-10G A656015-500G
103840-418EA 6 USD
103840-418 76844-604 76844-606 77293-054 77293-056 77293-058
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 98%
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane)

Precaución: for Research Use Only

Formula: C₂₀H₁₇ClO
MW: 308.8 g/mol
Melting Pt: 122…124 °C
Flash Pt: 208.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000814
CAS Number: 14470-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR