(Bromomethyl)triphenylphosphonium bromide 95%
Supplier: Ambeed
|
Warning
|
A200616-1G
A200616-25G
A200616-5G
A200616-100G
A200616-10G
A200616-500G
103817-222EA
7
USD
103817-222
103847-854
77085-912
77085-914
77085-916
77139-324
77139-326
77139-328
(Bromomethyl)triphenylphosphonium bromide 95%
(Bromomethyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₁₉H₁₇Br₂P MW: 436.1200 g/mol Melting Pt: 234…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011864 CAS Number: 1034-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...