(Aminomethyl)phosphonic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(Aminomethyl)phosphonic Acid, AMPA
A360059-1G
A360059-250MG
A360059-5G
103826-244EA
15
USD
103826-244
103837-252
76840-452
77142-290
77306-706
(Aminomethyl)phosphonic acid 97%
(Aminomethyl)phosphonic acid
Precaución: for Research Use Only
Formula:
CH₆NO₃P Boiling Pt: 358 °C (760 mm Hg) Melting Pt: 300 °C Flash Pt: 170.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008105 CAS Number: 1066-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...