Allyl phenyl ether 97%

Supplier: Ambeed
Warning

Synonyms: (Allyloxy)benzene

A121524-10G A121524-5G
76688-134EA 10.61 USD
76688-134 77312-206
Allyl phenyl ether 97%
Allyl phenyl ether

Precaución: for Research Use Only

Formula: H₂CCHCH₂OC₆H₅
MW: 134.18 g/mol
Boiling Pt: 192.4 °C (760 mm Hg)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008644
CAS Number: 1746-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR