(9H-Fluoren-9-yl)methyl 2-oxoethylcarbamate 96%

Supplier: Ambeed
Warning

A438720-25G A438720-5G A438720-10G A438720-1G
103827-636EA 39 USD
103827-636 103838-700 76842-446 76842-448 77143-354 77307-302
(9H-Fluoren-9-yl)methyl 2-oxoethylcarbamate 96%
(9H-Fluoren-9-yl)methyl 2-oxoethylcarbamate

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₃
MW: 281.3 g/mol
Melting Pt: 140…143 °C
Storage Temperature: Freezer
MDL Number: MFCD11519176
CAS Number: 156939-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR