(9H-Fluoren-9-yl)methyl (2-bromoethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Fmoc-amino)ethyl bromide

A347201-1G A347201-5G A347201-10G A347201-250MG
103825-962EA 9 USD
103825-962 103836-948 76840-024 76840-026 77251-492 77251-494
(9H-Fluoren-9-yl)methyl (2-bromoethyl)carbamate 95%
(9H-Fluoren-9-yl)methyl (2-bromoethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₁₆BrNO₂
MW: 346.22 g/mol
Boiling Pt: 491.9 °C (760 mm Hg)
Melting Pt: 122…124 °C
Flash Pt: 251.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797866
CAS Number: 340187-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR