9,9-Dimethylfluoren-2-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (9,9-Dimethyl-9H-fluoren-2-yl)boronic acid

A321829-1G A321829-100G A321829-10G A321829-250MG A321829-25G A321829-5G
103825-292EA 6 USD
103825-292 103836-226 76839-140 76978-082 76978-084 76978-086 77306-254 77306-256
9,9-Dimethylfluoren-2-boronic acid 97%
9,9-Dimethylfluoren-2-boronic acid

Precaución: For research use only

Formula: C₁₅H₁₅BO₂
MW: 238.09 g/mol
Boiling Pt: 436.4 °C (760 mm Hg)
Flash Pt: 217.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08704227
CAS Number: 333432-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR