(6R,7R)-4-Methoxybenzyl-7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
ACLE HCL, (6R,7R)-4-Methoxybenzyl 7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride
A102740-250MG
A102740-25G
A102740-1G
A102740-5G
103796-424EA
26
USD
103796-424
103822-120
76780-572
76780-574
77308-990
77308-992
(6R,7R)-4-Methoxybenzyl-7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride 95%
(6R,7R)-4-Methoxybenzyl-7-amino-3-(chloromethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₆H₁₈Cl₂N₂O₄S MW: 405.2961 g/mol Melting Pt: 155…157 °C Flash Pt: 317.4 °C Storage Temperature: Freezer |
MDL Number:
MFCD00191255 CAS Number: 113479-65-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...