2-Methyl-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Methylpyridin-3-yl)boronic acid

A216887-1G A216887-5G A216887-250MG
103818-586EA 14 USD
103818-586 103849-312 77087-918 77087-920 77165-122
2-Methyl-5-pyridineboronic acid 98%
2-Methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₂
MW: 136.94 g/mol
Flash Pt: 132 °C
Storage Temperature: Freezer
MDL Number: MFCD06801736
CAS Number: 659742-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR