(6-Methylpyridin-2-yl)methanamine 97%
Supplier: Ambeed
|
Warning
|
A245946-1G
A245946-100MG
A245946-10G
A245946-250MG
103811-678EA
20
USD
103811-678
103851-508
77090-782
77165-886
77165-888
77165-890
(6-Methylpyridin-2-yl)methanamine 97%
(6-Methylpyridin-2-yl)methanamine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂ Boiling Pt: 198.6 °C (760 mm Hg) Flash Pt: 94.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05864530 CAS Number: 6627-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...