2-Methoxy-5-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (6-Methoxypyridin-3-yl)boronic acid

A130822-100G A130822-10G A130822-250MG A130822-1G A130822-25G A130822-5G
103801-482EA 6 USD
103801-482 77109-224 77109-226 77109-228 77313-948 77313-950 77313-952
2-Methoxy-5-pyridineboronic acid 97%
2-Methoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
MW: 152.95 g/mol
Boiling Pt: 313.2 °C (760 mm Hg)
Melting Pt: 135…140 °C
Flash Pt: 143.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093044
CAS Number: 163105-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR