2-Isopropoxy-5-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (6-Isopropoxypyridin-3-yl)boronic acid, 2-Isoproxypyridine-5-boronic acid

A178432-1G A178432-5G A178432-250MG
103807-328EA 41 USD
103807-328 103845-668 77083-092 77083-094 77667-474
2-Isopropoxy-5-pyridineboronic acid 97%
2-Isopropoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181.00 g/mol
Boiling Pt: 325.5 °C (760 mm Hg)
Flash Pt: 150.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368870
CAS Number: 870521-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR