6-Hydroxy-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Hydroxypyridin-3-yl)boronic acid

A220650-100MG A220650-250MG
103818-868EA 26 USD
103818-868 103849-750 77030-804 77568-128
6-Hydroxy-3-pyridineboronic acid 98%
6-Hydroxy-3-pyridineboronic acid

Precaución: For research use only.

Formula: C₅H₆BNO₃
MW: 138.917 g/mol
Boiling Pt: 500.8 °C (760 mm Hg)
Flash Pt: 256.671 °C
Storage Temperature: Freezer
MDL Number: MFCD09907979
CAS Number: 903899-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR