(6-Fluoro-3-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: (6-Fluoropyridin-3-yl)methanol

A378773-1G A378773-5G A378773-100MG A378773-250MG
103826-632EA 24 USD
103826-632 103837-666 76840-978 76840-980 76978-820 76978-822
(6-Fluoro-3-pyridinyl)methanol 97%
(6-Fluoro-3-pyridinyl)methanol

Precaución: For research use only

Formula: C₆H₆FNO
MW: 127.1200 g/mol
Boiling Pt: 256.4 °C (760 mm Hg)
Flash Pt: 108.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095186
CAS Number: 39891-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR