(6-Fluoropyridin-3-yl)methylamine 97%

Supplier: Ambeed
Danger

Synonyms: (6-Fluoropyridin-3-yl)methanamine

A527734-100MG A527734-250MG
103828-548EA 101 USD
103828-548 103839-660 76980-050 77673-400
(6-Fluoropyridin-3-yl)methylamine 97%
(6-Fluoropyridin-3-yl)methylamine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.132 g/mol
Boiling Pt: 231.9 °C (760 mm Hg)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100747
CAS Number: 205744-17-8
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR