6-Fluoropyridine-2-boronic acid 95%
Supplier: Ambeed
|
Warning
|
A578238-100MG
A578238-250MG
A578238-1G
A578238-5G
103828-848EA
74
USD
103828-848
103839-998
76790-390
76790-392
77307-862
77307-864
6-Fluoropyridine-2-boronic acid 95%
Precaución: for Research Use Only
Formula:
C₅H₅BFNO₂ Storage Temperature: Freezer |
MDL Number:
MFCD08705713 CAS Number: 916176-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...