2-Fluoro-3-methyl-5-pyridineboronic acid 98%
Supplier: Ambeed
|
Warning
|
A244226-100MG
A244226-10G
A244226-250MG
A244226-1G
A244226-25G
A244226-5G
103811-572EA
12
USD
103811-572
103851-404
76786-638
76786-640
76786-642
77304-310
77304-312
77304-314
2-Fluoro-3-methyl-5-pyridineboronic acid 98%
2-Fluoro-3-methyl-5-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₆H₇BFNO₂ Melting Pt: 161…164 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03411562 CAS Number: 904326-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...