(6-Cyclopropylpyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A190391-100MG A190391-250MG A190391-1G
103816-292EA 388 USD
103816-292 76974-822 76974-824 77084-690
(6-Cyclopropylpyridin-3-yl)boronic acid 97%
(6-Cyclopropylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₂
MW: 162.982 g/mol
Boiling Pt: 353 °C (760 mm Hg)
Flash Pt: 167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18255555
CAS Number: 1253055-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR