2-Cyanopyridine-5-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (6-Cyanopyridin-3-yl)boronic acid

A190001-250MG A190001-1G A190001-5G A190001-10G A190001-25G
103816-248EA 80 USD
103816-248 103846-842 76873-022 77084-634 77084-636 77883-407 77883-408
2-Cyanopyridine-5-boronic acid 97%
2-Cyanopyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BN₂O₂
MW: 147.927 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696633
CAS Number: 1011722-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR