5-(Aminomethyl)-2-chloropyridine 96%

Supplier: Ambeed
Danger

Synonyms: (6-Chloropyridin-3-yl)methanamine

A444779-10G A444779-1G A444779-5G A444779-25G
103827-716EA 11 USD
103827-716 103838-784 76829-680 76842-564 76842-566 77307-328
5-(Aminomethyl)-2-chloropyridine 96%
5-(Aminomethyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 28…34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00673153
CAS Number: 97004-04-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR