2-Chloro-5-pyridineboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloropyridine-5-boronic acid, 2-Chloro-5-pyridinylboronic acid, 6-Chloro-3-pyridinylboronic acid, (6-Chloropyridin-3-yl)boronic acid, 6-Chloropyridine-3-boronic acid
A180503-25G
A180503-1G
A180503-10G
A180503-500G
103807-542EA
5
USD
103807-542
103845-898
77083-394
77301-674
77593-152
77683-449
2-Chloro-5-pyridineboronic acid 97%
2-Chloro-5-pyridineboronic acid
Precaución: For research use only.
Formula:
C₅H₅BClNO₂ MW: 157.36 g/mol Boiling Pt: 336.9 °C (760 mm Hg) Melting Pt: 165 °C Flash Pt: 157.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094998 CAS Number: 444120-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...