(6-Chloropyrazin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A282230-100MG A282230-10G A282230-250MG A282230-1G A282230-5G
103813-588EA 45 USD
103813-588 103853-540 76875-326 76875-328 76875-330 77305-428 77305-430
(6-Chloropyrazin-2-yl)methanol 97%
(6-Chloropyrazin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.5590 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15528712
CAS Number: 1240602-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR