(6-Chloropyrazin-2-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (6-Chloropyrazin-2-yl)methanamine hydrochloride

A312260-100MG A312260-250MG A312260-1G
103814-692EA 374 USD
103814-692 103835-882 76988-994 76988-996 77306-072
(6-Chloropyrazin-2-yl)methanamine hydrochloride 95%
(6-Chloropyrazin-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₆ClN₃·ClH
MW: 180.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20327986
CAS Number: 1357945-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR