(6-Chloro-5-(trifluoromethyl)pyridin-3-yl)methanol 97%

Supplier: Ambeed
Warning

A166626-100MG A166626-250MG A166626-1G
103806-084EA 190 USD
103806-084 76824-330 76824-332 77320-622
(6-Chloro-5-(trifluoromethyl)pyridin-3-yl)methanol 97%
(6-Chloro-5-(trifluoromethyl)pyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
Boiling Pt: 269.4±35.0 °C (760 mm Hg)
Flash Pt: 116.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12024295
CAS Number: 1113049-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR