(6-Bromo-3-pyridinyl)methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol
A362046-1G
A362046-25G
A362046-100G
A362046-10G
A362046-250MG
103826-298EA
13
USD
103826-298
103837-312
76840-526
76840-528
77003-358
77003-360
77003-362
(6-Bromo-3-pyridinyl)methanol 97%
(6-Bromo-3-pyridinyl)methanol
Precaución: for Research Use Only
Formula:
C₆H₆BrNO MW: 188.02 g/mol Melting Pt: 46…50 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD04974508 CAS Number: 122306-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...