5-(Aminomethyl)-2-bromopyridine 95%
Supplier: Ambeed
|
Warning
|
A109624-100MG
A109624-250MG
103797-536EA
36
USD
103797-536
103823-336
76687-452
76687-454
5-(Aminomethyl)-2-bromopyridine 95%
5-(Aminomethyl)-2-bromopyridine
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂ Boiling Pt: 290.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06738744 CAS Number: 120740-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...