2-Bromo-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

A113198-100G A113198-10G A113198-250MG A113198-25G A113198-1G A113198-5G
103798-186EA 8 USD
103798-186 103824-024 77009-036 77009-038 77009-040 77009-042 77297-230 77310-848
2-Bromo-5-pyridineboronic acid 98%
2-Bromo-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BBrNO₂
Boiling Pt: 358.2 °C (760 mm Hg)
Melting Pt: 161…166 °C
Flash Pt: 170.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03411558
CAS Number: 223463-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR