(6-Bromo-2-pyridinyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: (6-Bromopyridin-2-yl)methanol, 2-Bromo-6-pyridinemethanol, 2-Bromo-6-(hydroxymethyl)pyridine

A119297-10G A119297-250MG A119297-25G A119297-1G A119297-5G A119297-100G
103799-746EA 9 USD
103799-746 77284-744 77284-746 77284-748 77311-868 77311-870 77571-152
(6-Bromo-2-pyridinyl)methanol 98%
(6-Bromo-2-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 288.7 °C (760 mm Hg)
Melting Pt: 34…39 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04039884
CAS Number: 33674-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR