6-Bromo-5-methyl-3-pyridineboronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-5-methylopyridine-3-boronic acid, 6-Bromo-5-methyl-3-pyridinylboronic acid, 2-Bromo-3-methylpyridine-5-boronic acid, (6-Bromo-5-methylpyridin-3-yl)boronic acid
A216967-1G
A216967-5G
103818-592EA
16
USD
103818-592
103849-318
77087-932
77087-934
6-Bromo-5-methyl-3-pyridineboronic acid 95%
6-Bromo-5-methyl-3-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₆H₇BBrNO₂ MW: 215.84 g/mol Boiling Pt: 378.9 °C (760 mm Hg) Melting Pt: 204 °C Flash Pt: 183 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08274468 CAS Number: 1003043-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...