2-Amino-5-hydroxymethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-pyridinemethanol, 6-Amino-3-pyridinemethanol, 2-Amino-5-(hydroxymethyl)pyridine, 2-Aminopyridine-5-methanol, (6-Aminopyridin-3-yl)methanol, (6-Amino-3-pyridyl)methanol

A127666-10G A127666-250MG A127666-1G A127666-25G A127666-5G
103800-998EA 35 USD
103800-998 77136-798 77136-800 77313-252 77313-254 77313-256
2-Amino-5-hydroxymethylpyridine 98%
2-Amino-5-hydroxymethylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 320.2 °C (760 mm Hg)
Melting Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659089
CAS Number: 113293-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR