(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%

Supplier: Ambeed
Warning

A201783-1G A201783-10G A201783-250MG A201783-5G
103817-332EA 30 USD
103817-332 103847-972 77086-056 77139-346 77139-348 77302-782
(6-(Trifluoromethyl)pyridin-2-yl)methanol 98%
(6-(Trifluoromethyl)pyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
Boiling Pt: 191.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11100723
CAS Number: 131747-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR