(6-(Trifluoromethoxy)pyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A257503-100MG A257503-250MG A257503-1G A257503-25MG
103792-506EA 83 USD
103792-506 103812-310 76786-898 76786-900 77304-706 77304-708
(6-(Trifluoromethoxy)pyridin-3-yl)boronic acid 97%
(6-(Trifluoromethoxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₃NO₃
Boiling Pt: 259.7±50.0 °C (760 mm Hg)
Flash Pt: 110.9 °C
Storage Temperature: Freezer
MDL Number: MFCD07374890
CAS Number: 1008140-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR