(6-(Pyrrolidin-1-yl)pyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Pyrrolidin-1-yl)pyridine-3-boronic acid

A567056-10G A567056-250MG A567056-1G A567056-5G
103828-782EA 44 USD
103828-782 103839-924 76819-166 76819-168 76844-042 76844-044
(6-(Pyrrolidin-1-yl)pyridin-3-yl)boronic acid 97%
(6-(Pyrrolidin-1-yl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BN₂O₂
Boiling Pt: 421.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06801704
CAS Number: 1150114-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR