(6-(4-Methylpiperazin-1-yl)pyridin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-(4-Methylpiperazin-1-yl)pyridine-3-boronic acid

A256090-100MG A256090-250MG
103812-238EA 58 USD
103812-238 103852-100 77031-692 77031-694
(6-(4-Methylpiperazin-1-yl)pyridin-3-yl)boronic acid 95%
(6-(4-Methylpiperazin-1-yl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆BN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09907981
CAS Number: 936353-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR