[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butoxycarbonylamino)-pyridine-5-boronic acid, (6-((tert-Butoxycarbonyl)amino)pyridin-3-yl)boronic acid

A178048-100MG A178048-250MG A178048-1G A178048-5G
103807-296EA 99 USD
103807-296 103845-636 77029-320 77029-322 77083-034 77083-036
[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid 98%
[6-[(tert-Butoxycarbonyl)amino]pyridin-3-yl]boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD11975411
CAS Number: 883231-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR