(5-Nitropyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A134290-100MG A134290-250MG A134290-5G
103801-996EA 159 USD
103801-996 77285-422 77285-424 77298-708
(5-Nitropyridin-2-yl)methanol 97%
(5-Nitropyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
Boiling Pt: 327.6 °C (760 mm Hg)
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD07368186
CAS Number: 36625-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR