(5-Methylpyridin-2-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A119628-250MG A119628-5G
103799-804EA 115 USD
103799-804 76983-172 77297-706
(5-Methylpyridin-2-yl)methanamine hydrochloride 95%
(5-Methylpyridin-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD20731727
CAS Number: 161647-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR