5-Methyl-2-furanmethanol 95%

Supplier: Ambeed
Warning

Synonyms: (5-Methylfuran-2-yl)methanol, 2-(Hydroxymethyl)-5-methylfuran

A109518-100G A109518-10G A109518-25G A109518-1G A109518-5G
103797-516EA 21 USD
103797-516 103823-314 77135-914 77135-916 77135-918 77310-132 77310-134
5-Methyl-2-furanmethanol 95%
5-Methyl-2-furanmethanol

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.128 g/mol
Boiling Pt: 178.5 °C (760 mm Hg)
Flash Pt: 61.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130134
CAS Number: 3857-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR