(5-Methyl-1H-pyrazol-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A126737-100MG A126737-250MG
103800-858EA 122 USD
103800-858 77571-586 77592-460
(5-Methyl-1H-pyrazol-3-yl)boronic acid 97%
(5-Methyl-1H-pyrazol-3-yl)boronic acid

Precaución: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₄H₇BN₂O₂
MW: 125.92 g/mol
Storage Temperature: Freezer
MDL Number: MFCD14636621
CAS Number: 1163248-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR