(5-Methoxypyridin-2-yl)methanol 98%

Supplier: Ambeed
Warning

A316921-1G A316921-25G A316921-5G A316921-100MG A316921-10G A316921-250MG
103825-122EA 18 USD
103825-122 103836-054 76838-898 76838-900 76838-902 77141-738 77141-740 77141-742
(5-Methoxypyridin-2-yl)methanol 98%
(5-Methoxypyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Boiling Pt: 262 °C (760 mm Hg)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD08235020
CAS Number: 127978-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR