5-Fluoro-3-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid

A113732-10G A113732-250MG A113732-25G A113732-5G A113732-1G
103798-280EA 13 USD
103798-280 103824-124 77009-068 77009-070 77009-072 77297-264 77310-936
5-Fluoro-3-pyridineboronic acid 95%
5-Fluoro-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Melting Pt: 233…236 °C
Flash Pt: 130.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07368243
CAS Number: 872041-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR